COMH4211AA80 Combi-Blocks, Inc.

tert-Butyl 2-(4-amino-1h-pyrazol-1-yl)-2-methylpropanoate

  • MDL number MFCD30180417
  • Cas Number : 1462287-54-2
  • Molecular Weight : 225.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0796

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0796
Availability details for
Quantity
  • Availability
  • Add to Cart