COMH4211AA81 Combi-Blocks, Inc.

1-Isobutyl-1,2,3,4-tetrahydroquinolin-7-amine

  • MDL number MFCD11610020
  • Cas Number : 927684-32-0
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0797

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0797
Availability details for
Quantity
  • Availability
  • Add to Cart