COMH4211AA83 Combi-Blocks, Inc.

(Cyclopropylmethyl)(3-phenylpropyl)amine

  • MDL number MFCD11150466
  • Cas Number : 1020979-64-9
  • Molecular Weight : 189.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0800

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0800
Availability details for
Quantity
  • Availability
  • Add to Cart