COMH4211AA84 Combi-Blocks, Inc.

4-Amino-bicyclo[2.2.2]octane-1-carboxylic acid cyclopropylamide, hydrochloride

  • MDL number MFCD31539554
  • Cas Number : 2206607-70-5
  • Molecular Weight : 244.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0803

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0803
Availability details for
Quantity
  • Availability
  • Add to Cart