COMH4211AA85 Combi-Blocks, Inc.

Benzenemethanamine, 2-(2-methoxyethoxy)-alpha-methyl-

  • MDL number MFCD09813286
  • Cas Number : 1016735-05-9
  • Molecular Weight : 195.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0804

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0804
Availability details for
Quantity
  • Availability
  • Add to Cart