COMH4211AA87 Combi-Blocks, Inc.

5-Bromo-n-methyl-2-(2,2,2-trifluoro-ethoxy)-benzamide

  • MDL number MFCD31628793
  • Molecular Weight : 312.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0806

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0806
Availability details for
Quantity
  • Availability
  • Add to Cart