COMH4211AA88 Combi-Blocks, Inc.

4-(1H-1,2,4-Triazol-1-yl)butan-1-ol

  • MDL number MFCD12186771
  • Cas Number : 1153126-19-2
  • Molecular Weight : 141.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-(1H-1,2,4-triazol-1-yl)-1-butanol
Inchi Code 1S/C6H11N3O/c10-4-2-1-3-9-6-7-5-8-9/h5-6,10H,1-4H2
Inchi Key WMOLRTKCXVTIFO-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0807

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0807
Availability details for
Quantity
  • Availability
  • Add to Cart