COMH4211AA89 Combi-Blocks, Inc.

1-[4-(4-Methoxy-benzyloxy)-3-nitro-phenyl]-ethanone

  • MDL number MFCD31539517
  • Cas Number : 2204588-03-2
  • Molecular Weight : 301.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0811

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0811
Availability details for
Quantity
  • Availability
  • Add to Cart