COMH4211AA8A Combi-Blocks, Inc.

4'-Formyl-3-biphenylcarboxamide

  • MDL number MFCD31613782
  • Cas Number : 1007579-69-2
  • Molecular Weight : 225.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0812

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0812
Availability details for
Quantity
  • Availability
  • Add to Cart