COMH4211AA8E Combi-Blocks, Inc.

N-Methyl-n-propyl-isophthalamic acid

  • MDL number MFCD12173501
  • Cas Number : 1154356-99-6
  • Molecular Weight : 221.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0820

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0820
Availability details for
Quantity
  • Availability
  • Add to Cart