COMH4211AA8F Combi-Blocks, Inc.

1H-Pyrazole, 4-iodo-1-[2-(phenylmethoxy)ethyl]-

  • MDL number MFCD30483554
  • Cas Number : 1562305-17-2
  • Molecular Weight : 328.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0821

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0821
Availability details for
Quantity
  • Availability
  • Add to Cart