COMH4211AA90 Combi-Blocks, Inc.

4-[(3,4-Dichlorophenoxy)methyl]piperidine

  • MDL number MFCD08687537
  • Cas Number : 790205-11-7
  • Molecular Weight : 260.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0822

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0822
Availability details for
Quantity
  • Availability
  • Add to Cart