COMH4211AA91 Combi-Blocks, Inc.

3-(3-Bromo-phenyl)-propionic acid tert-butyl ester

  • MDL number MFCD22697141
  • Cas Number : 277331-50-7
  • Molecular Weight : 285.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0824

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0824
Availability details for
Quantity
  • Availability
  • Add to Cart