COMH4211AA92 Combi-Blocks, Inc.

4-Bromo-2-(2,2-difluoro-ethoxy)-benzoic acid

  • MDL number MFCD16108709
  • Cas Number : 1248672-38-9
  • Molecular Weight : 281.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0826

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0826
Availability details for
Quantity
  • Availability
  • Add to Cart