COMH4211AA94 Combi-Blocks, Inc.

1-(2-Bromo-5-nitro-benzyl)-4-ethyl-piperazine

  • MDL number MFCD31437589
  • Cas Number : 2138286-73-2
  • Molecular Weight : 328.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0830

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0830
Availability details for
Quantity
  • Availability
  • Add to Cart