COMH4211AA95 Combi-Blocks, Inc.

Bis-(2-chloro-ethyl)-(4-methoxy-benzyl)-amine

  • MDL number MFCD19332725
  • Cas Number : 91561-99-8
  • Molecular Weight : 262.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N,N-bis(2-chloroethyl)-N-(4-methoxybenzyl)amine
Inchi Code 1S/C12H17Cl2NO/c1-16-12-4-2-11(3-5-12)10-15(8-6-13)9-7-14/h2-5H,6-10H2,1H3
Inchi Key XVEHBTQWYNXBED-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0831

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0831
Availability details for
Quantity
  • Availability
  • Add to Cart