COMH4211AA96 Combi-Blocks, Inc.

(4-Amino-2-trifluoromethylphenyl)morpholin-4-yl-methanone

  • MDL number MFCD30486319
  • Cas Number : 919278-28-7
  • Molecular Weight : 274.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0836

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0836
Availability details for
Quantity
  • Availability
  • Add to Cart