COMH4211AA97 Combi-Blocks, Inc.

3-(2-Fluoro-ethoxy)-benzylamine

  • MDL number MFCD21647073
  • Cas Number : 1037078-87-7
  • Molecular Weight : 169.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0839

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0839
Availability details for
Quantity
  • Availability
  • Add to Cart