COMH4211AA9A Combi-Blocks, Inc.

Benzyl 2-(4-oxopiperidin-1-yl)acetate

  • MDL number MFCD07371645
  • Cas Number : 364056-14-4
  • Molecular Weight : 247.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0849

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0849
Availability details for
Quantity
  • Availability
  • Add to Cart