COMH4211AA9C Combi-Blocks, Inc.

3-(4-Amino-piperidin-1-yl)-2,2-dimethyl-propionic acid methyl ester hydrochloride

  • MDL number MFCD31557853
  • Cas Number : 2204559-95-3
  • Molecular Weight : 250.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0854

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0854
Availability details for
Quantity
  • Availability
  • Add to Cart