COMH4211AAA1 Combi-Blocks, Inc.

1-(4-Cyclopropylmethoxy-3-fluoro-phenyl)-ethanone

  • MDL number MFCD11136950
  • Cas Number : 1019626-67-5
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0879

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0879
Availability details for
Quantity
  • Availability
  • Add to Cart