COMH4211AAA2 Combi-Blocks, Inc.

N-(2-Phenylethyl)pentan-1-amine

  • MDL number MFCD08756384
  • Cas Number : 80377-02-2
  • Molecular Weight : 191.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0881

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0881
Availability details for
Quantity
  • Availability
  • Add to Cart