COMH4211AAA3 Combi-Blocks, Inc.

[(2-Bromo-4-chlorophenyl)methyl](methyl)amine

  • MDL number MFCD24119270
  • Cas Number : 900571-24-6
  • Molecular Weight : 234.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0882

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0882
Availability details for
Quantity
  • Availability
  • Add to Cart