COMH4211AAA4 Combi-Blocks, Inc.

2-(4-Bromo-2,6-difluoro-phenoxy)-2-methyl-propionic acid ethyl ester

  • MDL number MFCD31628747
  • Cas Number : 1203955-55-8
  • Molecular Weight : 323.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-(4-bromo-2,6-difluorophenoxy)-2-methylpropanoate
Inchi Code 1S/C12H13BrF2O3/c1-4-17-11(16)12(2,3)18-10-8(14)5-7(13)6-9(10)15/h5-6H,4H2,1-3H3
Inchi Key KNVIWTRUHULZRI-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0886

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0886
Availability details for
Quantity
  • Availability
  • Add to Cart