COMH4211AAA7 Combi-Blocks, Inc.

(4-Chloro-2-fluoro-benzyl)-cyclopropyl-amine hydrochloride

  • MDL number MFCD31613868
  • Cas Number : 2270905-92-3
  • Molecular Weight : 236.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(4-chloro-2-fluorobenzyl)cyclopropanamine hydrochloride
Inchi Code 1S/C10H11ClFN.ClH/c11-8-2-1-7(10(12)5-8)6-13-9-3-4-9;/h1-2,5,9,13H,3-4,6H2;1H
Inchi Key ZMAJSOBVMKEXEL-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0894

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0894
Availability details for
Quantity
  • Availability
  • Add to Cart