COMH4211AAA8 Combi-Blocks, Inc.

Phenyl-[4-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenoxy]-acetic acid methyl ester

  • MDL number MFCD31583133
  • Cas Number : 1029439-20-0
  • Molecular Weight : 368.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0896

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0896
Availability details for
Quantity
  • Availability
  • Add to Cart