COMH4211AAA9 Combi-Blocks, Inc.

5-Formyl-2-(1h-1,2,4-triazol-1-yl)benzonitrile

  • MDL number MFCD24678170
  • Cas Number : 1007315-95-8
  • Molecular Weight : 198.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0900

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0900
Availability details for
Quantity
  • Availability
  • Add to Cart