COMH4211AAAC Combi-Blocks, Inc.

1-(2-Fluoro-4-methyl-phenyl)-ethylamine hydrochloride

  • MDL number MFCD31539568
  • Cas Number : 2206265-54-3
  • Molecular Weight : 189.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0908

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0908
Availability details for
Quantity
  • Availability
  • Add to Cart