COMH4211AAAD Combi-Blocks, Inc.

3-Cyano-4-isobutoxybenzothioamide

  • MDL number MFCD13185967
  • Cas Number : 163597-57-7
  • Molecular Weight : 234.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-cyano-4-isobutoxybenzothioamide
Inchi Code 1S/C12H14N2OS/c1-8(2)7-15-11-4-3-9(12(14)16)5-10(11)6-13/h3-5,8H,7H2,1-2H3,(H2,14,16)
Inchi Key FMHRQJJWJQGSDR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0911

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0911
Availability details for
Quantity
  • Availability
  • Add to Cart