COMH4211AAAF Combi-Blocks, Inc.

L-Valine, n-[(2-propen-1-yloxy)carbonyl]-

  • MDL number MFCD02094428
  • Cas Number : 115491-96-8
  • Molecular Weight : 201.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0915

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0915
Availability details for
Quantity
  • Availability
  • Add to Cart