COMH4211AAB0 Combi-Blocks, Inc.

2-(1-(4-Methylbenzyl)piperidin-4-yl)ethan-1-ol

  • MDL number MFCD29993921
  • Cas Number : 185434-50-8
  • Molecular Weight : 233.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0917

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0917
Availability details for
Quantity
  • Availability
  • Add to Cart