COMH4211AAB1 Combi-Blocks, Inc.

6-Chloro-n-((tetrahydro-2h-pyran-4-yl)methyl)nicotinamide

  • MDL number MFCD19160058
  • Cas Number : 676626-73-6
  • Molecular Weight : 254.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0919

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0919
Availability details for
Quantity
  • Availability
  • Add to Cart