COMH4211AAB3 Combi-Blocks, Inc.

N-Hydroxy-(5-chloropyridine)-2-carbonimidoyl chloride

  • MDL number MFCD28660773
  • Cas Number : 1009562-96-2
  • Molecular Weight : 191.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0922

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0922
Availability details for
Quantity
  • Availability
  • Add to Cart