COMH4211AAB4 Combi-Blocks, Inc.

2-Chloro-n-isopropylnicotinamide

  • MDL number MFCD01356915
  • Cas Number : 56904-63-3
  • Molecular Weight : 198.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-N-isopropylnicotinamide
Inchi Code 1S/C9H11ClN2O/c1-6(2)12-9(13)7-4-3-5-11-8(7)10/h3-6H,1-2H3,(H,12,13)
Inchi Key VOZRJJMDZPIFHZ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0923

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0923
Availability details for
Quantity
  • Availability
  • Add to Cart