COMH4211AAB5 Combi-Blocks, Inc.

Ethyl-2-(5-bromo-2,4-difluoro-benzoyl)-3-[(4-methoxyphenyl)methylamino]prop-2-enoate

  • MDL number MFCD31579509
  • Cas Number : 2270915-01-8
  • Molecular Weight : 454.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0927

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0927
Availability details for
Quantity
  • Availability
  • Add to Cart