COMH4211AAB6 Combi-Blocks, Inc.

5-(4-Bromo-phenoxy)-2-nitro-benzoic acid methyl ester

  • MDL number MFCD31614108
  • Cas Number : 2270907-00-9
  • Molecular Weight : 352.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0930

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0930
Availability details for
Quantity
  • Availability
  • Add to Cart