COMH4211AAB8 Combi-Blocks, Inc.

4-Chloro-2-(1h-pyrazol-1-yl)benzaldehyde

  • MDL number MFCD30272979
  • Cas Number : 1446818-92-3
  • Molecular Weight : 206.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0933

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0933
Availability details for
Quantity
  • Availability
  • Add to Cart