COMH4211AABA Combi-Blocks, Inc.

Benzene, 1-(4-bromobutoxy)-4-(methylsulfonyl)-

  • MDL number MFCD12066160
  • Cas Number : 1157383-64-6
  • Molecular Weight : 307.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0937

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0937
Availability details for
Quantity
  • Availability
  • Add to Cart