COMH4211AAC0 Combi-Blocks, Inc.

2-Methyl-3-m-tolyl-propionic acid

  • MDL number MFCD09894702
  • Cas Number : 859192-05-5
  • Molecular Weight : 178.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0948

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0948
Availability details for
Quantity
  • Availability
  • Add to Cart