COMH4211AAC1 Combi-Blocks, Inc.

3-(4-Formylbenzamido)propanoic acid

  • MDL number MFCD25370394
  • Cas Number : 127560-07-0
  • Molecular Weight : 221.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0951

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0951
Availability details for
Quantity
  • Availability
  • Add to Cart