COMH4211AAC2 Combi-Blocks, Inc.

(3-Chloro-2-isopropoxy-benzyl)-methyl-amine

  • MDL number MFCD24611651
  • Cas Number : 894851-28-6
  • Molecular Weight : 213.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0952

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0952
Availability details for
Quantity
  • Availability
  • Add to Cart