COMH4211AAC3 Combi-Blocks, Inc.

2-(2,3-Dimethylphenoxy)ethan-1-ol

  • MDL number MFCD11613990
  • Cas Number : 83414-61-3
  • Molecular Weight : 166.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0953

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0953
Availability details for
Quantity
  • Availability
  • Add to Cart