COMH4211AAC5 Combi-Blocks, Inc.

3-Bromo-4-(ethylsulfanyl)aniline

  • MDL number MFCD16085614
  • Cas Number : 1250043-60-7
  • Molecular Weight : 232.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0957

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0957
Availability details for
Quantity
  • Availability
  • Add to Cart