COMH4211AAC7 Combi-Blocks, Inc.

4-(4,4-Difluoro-piperidin-1-yl)-butyric acid methyl ester

  • MDL number MFCD31579334
  • Cas Number : 1896964-16-1
  • Molecular Weight : 221.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0961

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0961
Availability details for
Quantity
  • Availability
  • Add to Cart