COMH4211AACA Combi-Blocks, Inc.

N-(5-Aminopentyl)benzenesulfonamide

  • MDL number MFCD20325366
  • Cas Number : 58885-30-6
  • Molecular Weight : 242.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0971

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0971
Availability details for
Quantity
  • Availability
  • Add to Cart