COMH4211AACB Combi-Blocks, Inc.

2-(3-Methoxy-benzyl)-butyric acid

  • MDL number MFCD09894731
  • Cas Number : 177362-57-1
  • Molecular Weight : 208.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0972

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0972
Availability details for
Quantity
  • Availability
  • Add to Cart