COMH4211AACC Combi-Blocks, Inc.

2-Fluoro-4-iodo-6-chloroaniline

  • MDL number MFCD20484389
  • Cas Number : 702640-40-2
  • Molecular Weight : 271.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0974

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0974
Availability details for
Quantity
  • Availability
  • Add to Cart