COMH4211AACD Combi-Blocks, Inc.

3-Amino-5-bromo-n-(prop-2-en-1-yl)benzamide

  • MDL number MFCD24125295
  • Cas Number : 1517751-02-8
  • Molecular Weight : 255.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0976

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0976
Availability details for
Quantity
  • Availability
  • Add to Cart