COMH4211AACE Combi-Blocks, Inc.

5-Bromo-2-isopropoxy-n-isopropyl-benzamide

  • MDL number MFCD31628787
  • Cas Number : 2270909-37-8
  • Molecular Weight : 300.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0984

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0984
Availability details for
Quantity
  • Availability
  • Add to Cart