COMH4211AACF Combi-Blocks, Inc.

5-Methyl-2-(prop-2-en-1-yloxy)benzoic acid

  • MDL number MFCD14536043
  • Cas Number : 1061592-06-0
  • Molecular Weight : 192.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0986

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-0986
Availability details for
Quantity
  • Availability
  • Add to Cart